Assay Details
| Gene Symbol: let-805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027469.3 | NP_001022640.1 | 20 - 21 | 11975 | 75 | |
| NM_001027470.3 | NP_001022641.1 | 20 - 21 | 11966 | 75 | ||
| NM_001306713.4 | NP_001293642.1 | 3 - 4 | 422 | 75 | ||
| GenBank mRNA | AF148953.1 | - | 20 - 21 | 11973 | 75 | |
| AF148954.1 | - | 20 - 21 | 11982 | 75 | ||
More Information
Set Membership: |
|

let-805
Genomic Map