Assay Details
| Gene Symbol: let-805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027470.3 | NP_001022641.1 | 25 - 26 | 13198 | 55 | |
| GenBank mRNA | AF148953.1 | - | 25 - 26 | 13205 | 55 | |
More Information
Set Membership: |
|

let-805
Genomic Map