Assay Details
| Gene Symbol: laf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267930.3 | NP_001254859.1 | 5 - 6 | 1981 | 151 | |
| GenBank mRNA | FJ348231.1 | - | 5 - 6 | 2003 | 151 | |
More Information
Set Membership: |
|

laf-1
Genomic Map