Assay Details
| Gene Symbol: daf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312987.3 | NP_001299916.1 | 11 | 2669 | 80 | |
| NM_001312988.3 | NP_001299917.1 | 9 - 10 | 2351 | 80 | ||
| NM_001312989.3 | NP_001299918.1 | 8 - 9 | 2264 | 80 | ||
| NM_001312990.3 | NP_001299919.1 | 1 - 2 | 62 | 80 | ||
| NM_065249.6 | NP_497650.4 | 11 - 12 | 2669 | 80 | ||
| NR_132448.1 | 10 - 11 | 2492 | 80 | |||
More Information
Set Membership: |
|

daf-2
Genomic Map