Assay Details
| Gene Symbol: daf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312987.3 | NP_001299916.1 | 15 | 3876 | 76 | |
| NM_001312988.3 | NP_001299917.1 | 12 - 13 | 3312 | 76 | ||
| NM_001312989.3 | NP_001299918.1 | 11 - 12 | 3225 | 76 | ||
| NM_001312990.3 | NP_001299919.1 | 4 - 5 | 1023 | 76 | ||
| NM_065249.6 | NP_497650.4 | 14 - 15 | 3630 | 76 | ||
| NR_132448.1 | 13 - 14 | 4640 | 76 | |||
More Information
Set Membership: |
|

daf-2
Genomic Map