Assay Details
| Gene Symbol: daf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312987.3 | NP_001299916.1 | 16 | 5409 | 74 | |
| NM_001312988.3 | NP_001299917.1 | 13 - 14 | 4845 | 74 | ||
| NM_001312989.3 | NP_001299918.1 | 12 - 13 | 4758 | 74 | ||
| NM_001312990.3 | NP_001299919.1 | 5 - 6 | 2556 | 74 | ||
| NM_001312991.3 | NP_001299920.1 | 1 - 2 | 1464 | 74 | ||
| NM_065249.6 | NP_497650.4 | 15 - 16 | 5163 | 74 | ||
| NR_132448.1 | 14 - 15 | 6173 | 74 | |||
More Information
Set Membership: |
|

daf-2
Genomic Map