Assay Details
| Gene Symbol: daf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312987.3 | NP_001299916.1 | 2 | 240 | 84 | |
| NM_065249.6 | NP_497650.4 | 2 - 3 | 240 | 84 | ||
| NR_132448.1 | 1 - 2 | 63 | 84 | |||
More Information
Set Membership: |
|

daf-2
Genomic Map