Assay Details
| Gene Symbol: mks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381800.1 | NP_001366806.1 | 8 - 9 | 1546 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mks-1| Gene Symbol: mks-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381800.1 | NP_001366806.1 | 8 - 9 | 1546 | 73 | |
Set Membership: |
|