Assay Details
| Gene Symbol: spe-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267943.3 | NP_001254872.1 | 3 - 4 | 787 | 121 | |
| NM_001267944.4 | NP_001254873.1 | 1 - 2 | 53 | 121 | ||
| GenBank mRNA | AJ276029.1 | - | 1 - 2 | 65 | 121 | |
| AY342006.1 | - | 1 - 2 | 83 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267944.4 transcripts.
Set Membership: |
|

spe-41
Genomic Map