Assay Details
| Gene Symbol: spe-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267943.3 | NP_001254872.1 | 4 - 5 | 885 | 134 | |
| NM_001267944.4 | NP_001254873.1 | 2 - 3 | 151 | 134 | ||
| GenBank mRNA | AJ276029.1 | - | 2 - 3 | 163 | 134 | |
| AY342006.1 | - | 2 - 3 | 181 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267944.4 transcripts.
Set Membership: |
|

spe-41
Genomic Map