Assay Details
| Gene Symbol: spe-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267943.3 | NP_001254872.1 | 7 - 8 | 1279 | 150 | |
| NM_001267944.4 | NP_001254873.1 | 5 - 6 | 542 | 150 | ||
| GenBank mRNA | AJ276029.1 | - | 5 - 6 | 557 | 150 | |
| AY342006.1 | - | 5 - 6 | 575 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267944.4 transcripts.
Set Membership: |
|

spe-41
Genomic Map