Assay Details
| Gene Symbol: spe-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267943.3 | NP_001254872.1 | 9 - 10 | 1690 | 96 | |
| NM_001267944.4 | NP_001254873.1 | 7 - 8 | 953 | 96 | ||
| GenBank mRNA | AJ276029.1 | - | 7 - 8 | 968 | 96 | |
| AY342006.1 | - | 7 - 8 | 986 | 96 | ||
More Information
Set Membership: |
|

spe-41
Genomic Map