Assay Details
| Gene Symbol: spe-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267943.3 | NP_001254872.1 | 10 - 11 | 1908 | 93 | |
| NM_001267944.4 | NP_001254873.1 | 8 - 9 | 1171 | 93 | ||
| GenBank mRNA | AJ276029.1 | - | 8 - 9 | 1186 | 93 | |
| AY342006.1 | - | 8 - 9 | 1204 | 93 | ||
More Information
Set Membership: |
|

spe-41
Genomic Map