Assay Details
| Gene Symbol: C34C12.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065316.5 | NP_497717.2 | 3 - 4 | 612 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C34C12.6| Gene Symbol: C34C12.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065316.5 | NP_497717.2 | 3 - 4 | 612 | 74 | |
Set Membership: |
|