Assay Details
| Gene Symbol: M01F1.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065321.8 | NP_497722.3 | 5 - 6 | 877 | 67 | |
| Gene Symbol: rpl-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381806.2 | NP_001366705.1 | 4 - 5 | 736 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381806.2 transcripts.
Set Membership: |
|

M01F1.3
Genomic Map