Assay Details
| Gene Symbol: npr-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065343.4 | NP_497744.2 | 6 - 7 | 950 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

npr-15| Gene Symbol: npr-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065343.4 | NP_497744.2 | 6 - 7 | 950 | 73 | |
Set Membership: |
|