Assay Details
| Gene Symbol: cyn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065344.5 | NP_497745.1 | 4 - 5 | 745 | 150 | |
| GenBank mRNA | U36581.1 | - | 4 - 5 | 738 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyn-9| Gene Symbol: cyn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065344.5 | NP_497745.1 | 4 - 5 | 745 | 150 | |
| GenBank mRNA | U36581.1 | - | 4 - 5 | 738 | 150 | |
Set Membership: |
|