Assay Details
| Gene Symbol: pdi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065345.7 | NP_497746.1 | 3 - 4 | 1288 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdi-1| Gene Symbol: pdi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065345.7 | NP_497746.1 | 3 - 4 | 1288 | 62 | |
Set Membership: |
|