Assay Details
| Gene Symbol: evl-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065355.5 | NP_497756.2 | 7 - 8 | 1571 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

evl-18| Gene Symbol: evl-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065355.5 | NP_497756.2 | 7 - 8 | 1571 | 98 | |
Set Membership: |
|