Assay Details
| Gene Symbol: mtm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027623.6 | NP_001022794.2 | 3 - 4 | 384 | 103 | |
| NM_065365.9 | NP_497766.3 | 3 - 4 | 372 | 103 | ||
| GenBank mRNA | DQ988041.1 | - | 3 - 4 | 233 | 103 | |
More Information
Set Membership: |
|

mtm-3
Genomic Map