Assay Details
| Gene Symbol: mtm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027623.6 | NP_001022794.2 | 5 - 6 | 1029 | 97 | |
| NM_065365.9 | NP_497766.3 | 5 - 6 | 1017 | 97 | ||
| GenBank mRNA | DQ988041.1 | - | 5 - 6 | 878 | 97 | |
More Information
Set Membership: |
|

mtm-3
Genomic Map