Assay Details
| Gene Symbol: mtm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027623.6 | NP_001022794.2 | 9 - 10 | 3200 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtm-3| Gene Symbol: mtm-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027623.6 | NP_001022794.2 | 9 - 10 | 3200 | 147 | |
Set Membership: |
|