Assay Details
| Gene Symbol: dpy-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065370.6 | NP_497771.1 | 12 - 13 | 4231 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dpy-27| Gene Symbol: dpy-27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065370.6 | NP_497771.1 | 12 - 13 | 4231 | 69 | |
Set Membership: |
|