Assay Details
| Gene Symbol: amx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065371.6 | NP_497772.2 | 4 - 5 | 826 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

amx-1| Gene Symbol: amx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065371.6 | NP_497772.2 | 4 - 5 | 826 | 100 | |
Set Membership: |
|