Assay Details
| Gene Symbol: rpl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026083.6 | NP_001021254.1 | 3 - 4 | 957 | 75 | |
| NM_001026084.7 | NP_001021255.1 | 3 - 4 | 798 | 75 | ||
| NM_001026085.5 | NP_001021256.1 | 3 - 4 | 648 | 75 | ||
| GenBank mRNA | Z69336.1 | - | 3 - 4 | 958 | 75 | |
More Information
Set Membership: |
|

rpl-3
Genomic Map