Assay Details
| Gene Symbol: tir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026080.5 | NP_001021251.2 | 3 - 4 | 992 | 76 | |
| NM_001312937.3 | NP_001299866.1 | 3 | 870 | 76 | ||
| NM_001381811.1 | NP_001368535.1 | 3 - 4 | 708 | 76 | ||
| NM_001381812.1 | NP_001367074.1 | 3 - 4 | 992 | 76 | ||
| NM_001384059.1 | 5 - 6 | 1017 | 76 | |||
| NM_065383.4 | NP_497784.1 | 5 - 6 | 1034 | 76 | ||
| GenBank mRNA | AY834227.1 | - | 3 - 4 | 872 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384059.1 transcripts.
Set Membership: |
|

tir-1
Genomic Map