Assay Details
| Gene Symbol: tir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026080.5 | NP_001021251.2 | 7 - 8 | 1831 | 67 | |
| NM_001312937.3 | NP_001299866.1 | 6 - 7 | 1709 | 67 | ||
| NM_001381811.1 | NP_001368535.1 | 7 - 8 | 1547 | 67 | ||
| NM_001381812.1 | NP_001367074.1 | 7 - 8 | 1831 | 67 | ||
| NM_001384059.1 | 9 - 10 | 1856 | 67 | |||
| NM_065383.4 | NP_497784.1 | 9 - 10 | 1873 | 67 | ||
| GenBank mRNA | AY834227.1 | - | 7 - 8 | 1711 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384059.1 transcripts.
Set Membership: |
|

tir-1
Genomic Map