Assay Details
| Gene Symbol: tir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026080.5 | NP_001021251.2 | 8 - 9 | 2861 | 109 | |
| NM_001312937.3 | NP_001299866.1 | 8 | 2739 | 109 | ||
| NM_001312939.3 | NP_001299868.1 | 1 - 2 | 825 | 109 | ||
| NM_001381811.1 | NP_001368535.1 | 8 - 9 | 2577 | 109 | ||
| NM_001381812.1 | NP_001367074.1 | 8 - 9 | 2861 | 109 | ||
| NM_001384059.1 | 10 - 11 | 2886 | 109 | |||
| NM_065383.4 | NP_497784.1 | 10 - 11 | 2903 | 109 | ||
| NM_065384.7 | NP_497785.1 | 2 - 3 | 1569 | 109 | ||
| GenBank mRNA | AY834227.1 | - | 8 - 9 | 2741 | 109 | |
More Information
Set Membership: |
|

tir-1
Genomic Map