Assay Details
| Gene Symbol: tir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026080.5 | NP_001021251.2 | 9 - 10 | 2977 | 62 | |
| NM_001381811.1 | NP_001368535.1 | 9 - 10 | 2693 | 62 | ||
| NM_001384059.1 | 11 - 12 | 3002 | 62 | |||
| NM_065384.7 | NP_497785.1 | 3 - 4 | 1685 | 62 | ||
More Information
Set Membership: |
|

tir-1
Genomic Map