Assay Details
| Gene Symbol: mmcm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065385.7 | NP_497786.2 | 3 - 4 | 1339 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mmcm-1| Gene Symbol: mmcm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065385.7 | NP_497786.2 | 3 - 4 | 1339 | 119 | |
Set Membership: |
|