Assay Details
| Gene Symbol: pat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065386.6 | NP_497787.1 | 6 - 7 | 2086 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pat-3| Gene Symbol: pat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065386.6 | NP_497787.1 | 6 - 7 | 2086 | 79 | |
Set Membership: |
|