Assay Details
| Gene Symbol: msi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384060.1 | 2 - 3 | 434 | 84 | ||
| GenBank mRNA | AB036738.1 | - | 2 - 3 | 434 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

msi-1| Gene Symbol: msi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384060.1 | 2 - 3 | 434 | 84 | ||
| GenBank mRNA | AB036738.1 | - | 2 - 3 | 434 | 84 | |
Set Membership: |
|