Assay Details
| Gene Symbol: rcq-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322778.4 | NP_001309439.1 | 1 - 2 | 139 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rcq-5| Gene Symbol: rcq-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322778.4 | NP_001309439.1 | 1 - 2 | 139 | 79 | |
Set Membership: |
|