Assay Details
| Gene Symbol: rnf-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322782.3 | NP_001309592.1 | 4 - 5 | 551 | 77 | |
| NM_001322783.3 | NP_001309594.1 | 2 - 3 | 185 | 77 | ||
| NM_001322784.3 | NP_001309593.1 | 1 - 2 | 236 | 77 | ||
| NM_065429.4 | NP_497830.1 | 3 - 4 | 602 | 77 | ||
More Information
Set Membership: |
|

rnf-5
Genomic Map