Assay Details
| Gene Symbol: rnf-121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065431.6 | NP_497832.2 | 3 - 4 | 399 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rnf-121| Gene Symbol: rnf-121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065431.6 | NP_497832.2 | 3 - 4 | 399 | 155 | |
Set Membership: |
|