Assay Details
| Gene Symbol: evl-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065464.8 | NP_497865.2 | 18 - 19 | 3599 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

evl-14| Gene Symbol: evl-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065464.8 | NP_497865.2 | 18 - 19 | 3599 | 104 | |
Set Membership: |
|