Assay Details
| Gene Symbol: cdk-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267987.3 | NP_001254916.1 | 6 - 7 | 766 | 105 | |
| NM_001381836.2 | NP_001367220.1 | 6 - 7 | 793 | 105 | ||
| NM_001381837.2 | NP_001367221.1 | 6 - 7 | 790 | 105 | ||
More Information
Set Membership: |
|

cdk-12
Genomic Map