Assay Details
| Gene Symbol: lcmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065474.4 | NP_497875.1 | 6 - 7 | 934 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lcmt-1| Gene Symbol: lcmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065474.4 | NP_497875.1 | 6 - 7 | 934 | 112 | |
Set Membership: |
|