Assay Details
| Gene Symbol: ckb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065478.4 | NP_497879.2 | 4 - 5 | 538 | 93 | |
| GenBank mRNA | AY158690.1 | - | 3 - 4 | 372 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ckb-1| Gene Symbol: ckb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065478.4 | NP_497879.2 | 4 - 5 | 538 | 93 | |
| GenBank mRNA | AY158690.1 | - | 3 - 4 | 372 | 93 | |
Set Membership: |
|