Assay Details
| Gene Symbol: ckb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065479.8 | NP_497880.2 | 4 - 5 | 368 | 100 | |
| GenBank mRNA | AY158691.1 | - | 2 - 3 | 276 | 100 | |
More Information
Set Membership: |
|

ckb-2
Genomic Map