Assay Details
| Gene Symbol: ckb-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065479.8 | NP_497880.2 | 8 - 9 | 970 | 124 | |
| GenBank mRNA | AY158691.1 | - | 6 - 7 | 878 | 124 | |
More Information
Set Membership: |
|

ckb-2
Genomic Map