Assay Details
| Gene Symbol: pals-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065481.8 | NP_497882.3 | 3 - 4 | 1814 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-26| Gene Symbol: pals-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065481.8 | NP_497882.3 | 3 - 4 | 1814 | 100 | |
Set Membership: |
|