Assay Details
| Gene Symbol: atf-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384076.1 | 5 - 6 | 1493 | 70 | ||
| NM_001384077.1 | 5 - 6 | 1633 | 70 | |||
| NM_065512.4 | NP_497913.1 | 6 - 7 | 1376 | 70 | ||
| NM_065513.5 | NP_497914.2 | 6 - 7 | 1367 | 70 | ||
More Information
Set Membership: |
|

atf-7
Genomic Map