Assay Details
| Gene Symbol: atf-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065513.5 | NP_497914.2 | 5 - 6 | 1255 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atf-7| Gene Symbol: atf-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065513.5 | NP_497914.2 | 5 - 6 | 1255 | 60 | |
Set Membership: |
|