Assay Details
| Gene Symbol: M88.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065521.4 | NP_497922.2 | 5 - 6 | 548 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

M88.4| Gene Symbol: M88.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065521.4 | NP_497922.2 | 5 - 6 | 548 | 103 | |
Set Membership: |
|