Assay Details
| Gene Symbol: pan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306693.3 | NP_001293622.1 | 1 - 2 | 133 | 159 | |
| NM_065523.8 | NP_497924.2 | 4 - 5 | 712 | 159 | ||
| NM_065524.5 | NP_497925.1 | 9 - 10 | 1654 | 159 | ||
More Information
Set Membership: |
|

pan-1
Genomic Map