Assay Details
| Gene Symbol: pan-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065523.8 | NP_497924.2 | 1 - 2 | 154 | 113 | |
| NM_065524.5 | NP_497925.1 | 6 - 7 | 1096 | 113 | ||
More Information
Set Membership: |
|

pan-1
Genomic Map