Assay Details
| Gene Symbol: metl-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065589.6 | NP_497990.1 | 2 - 3 | 302 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

metl-5| Gene Symbol: metl-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065589.6 | NP_497990.1 | 2 - 3 | 302 | 132 | |
Set Membership: |
|