Assay Details
| Gene Symbol: pal-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026038.6 | NP_001021209.1 | 2 - 3 | 262 | 79 | |
| NM_001026039.6 | NP_001021210.1 | 2 - 3 | 312 | 79 | ||
More Information
Set Membership: |
|

pal-1
Genomic Map