Assay Details
| Gene Symbol: F26F4.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065629.5 | NP_498030.1 | 4 - 5 | 481 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F26F4.12| Gene Symbol: F26F4.12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065629.5 | NP_498030.1 | 4 - 5 | 481 | 87 | |
Set Membership: |
|